Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Eisco™ Premium Stainless Steel Lab Jacks
Scissor style, premium stainless steel lab jacks used for regulating the height position of an object as a support apparatus.
Supplier: Eisco™ SSJCT20
Description
- Easy, fluid platform movement (with and without load) for precise height adjustments
- Single piece base enhances torsional rigidity for stabilization under load
- Made of high quality 304 stainless steel that is resistant to rust and corrosion
- Resistant to many chemicals and elements
- Can be autoclaved and chemically cleaned
- Features slip-proof rubber compression feet to avoid surface scratching
- Designed for use in biology, chemistry, physics, environmental, and biotechnology labs
Specifications
Stainless Steel | |
6.5 x 5 in. (Plate) | |
Slip Proof Rubber Feet | |
Autoclavable | |
General Laboratory Equipment |
11 in. (Max) | |
15.4 lb. | |
Scissor Jack | |
Regulating the Height Position of an Object as a Support Apparatus |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction