Learn More
Fisherbrand™ Stainless Steel Cleanroom Lab Table with Chemical Resistant Phenolic Resin Top - PROMO
FisherBrand™ Class 1 Cleanroom Lab Table features an adjustable height range with 2-inch increments and a 3 minute assembly
Description
- Grade 304 solid stainless steel frame
- No.4 brushed finish stainless with chemical resistant phenolic resin top
- Easy to clean
- Meets and exceeds SEFA
Specifications
Cleanroom Table | |
762 to 914 mm | |
36 in. | |
609 mm | |
30 to 36 in. (2 in. increments) | |
5,000 lb | |
Bench Frame, legs and top | |
Stainless Steel (304) |
2 in. | |
ANSI / BIFMA, SEFA | |
30 to 36 in. | |
Stainless | |
1219 mm | |
25 year unconditional manufacturer warranty | |
48 in. |