Learn More
Fine Science Tools Trephines for Micro Drill
Supplier: Fine Science Tools 1800423
Tip Diameter: 2.3mm/ Shaft Diameter: 2.3mm/ Alloy / Material: Stainless Steel/ Length: 44mm/ Autoclave Safe: Yes. These trephines with a side opening are ideal for clearing bone and other materials.