Learn More
Fine Science Tools Insect Pins, Tip: 0.03 mm, 4 cm, 100/pk
Supplier: Fine Science Tools 2600145
Insect Pins - Stainless Steel/Size 2/ Size: 2/ Tip Diameter: 0.03mm/ Rod Diameter: 0.45mm/ Alloy: Stainless Steel/ Length: 4cm/ Unit: 100