Learn More
Quanterix Inc SR-X SEALING OIL
Supplier: Quanterix Inc 102767
One bottle of Simoa oil for sealing the microwell arrays on the Quanterix SR-X. This quantity is sufficient for running approximately 50 plates (96