Learn More
Havels Sprotte needle 22G X 6" (150MM) With Introducer
Supplier: Havels INS22150
The Atraumatic Sprotte Spinal Needle for regional anesthesia and lumbar puncture reduces post-spinal headaches by a factor of ten! The conical point reduces dura trauma because it spreads dura fibers - it doesn't cut. The special opening allows CSF to flow freely and the rounded edges prevent coring.
This item is not returnable.
View return policy