Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ORS Nasco Sprayon™ Blast-Em™ Wasp and Hornet Killer

Supplier:  ORS Nasco 425S02085700

Catalog No. 19-313-648

Item Discontinued Due to product restrictions, please to purchase or view availability for this product.This item has been discontinued by the supplier.


Description

Description

  • Residual deposit will kill returning wasps and hornets (read label for proper indoor application)
  • Will not harm most plastics, including Noryl, ABS and Lexan
  • 98% knockdown, 100% kill within 24 hours
  • Dielectric strength: 33,000V
  • Sprays up to 20 ft. (6.1m)
    EPA-registered
  • Contains a high amount of petroleum distillates
  • Kills wasps, hornets, yellow jackets, flies, mosquitoes and flying moths
  • Aerosol spray, 16 oz. (473mL)

Specifications

Specifications

Wasp and Hornet Killer
Utility Meters and Boxes
20 oz.
Outside Surfaces of Screens, Doors and Window Frames, Porch Lighting Fixtures also for use in Outdoor Public Areas such as Golf Courses, Beaches, Picnic Grounds and Amphitheaters
0.59 L
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit