Learn More
Consolidated Plastics 32 Oz. Spraymaster
Supplier: Consolidated Plastics 41789
Chemical resistant Spraymaster spray bottles can be used with all types of cleaners, degreasers, and strippers. It sprays more than twice as much per stroke as standard chemical resistant sprayers. The opaque silver color protects light sensitive chemicals. A convenient "window-stripe" is available to view the liquid level on our Spraymaster spray bottle, and the levels can be measured with three graduation scales imprinted on the side.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.