Learn More
Grainger White/Clear Plastic Spray Bottle, 16 oz
Supplier: Grainger 5MN56
These spray bottles include a matching trigger sprayer to ensure compatibility. The trigger sprayer has an adjustable nozzle to change from a fine mist to a concentrated stream.