Learn More
WELCH ALLYN INC Spot Vital Signs 4400 Monitor
Supplier: WELCH ALLYN INC 44XTB
The Welch Allyn Spot 4400 Vital Signs Monitor is an automated, state-of-the-art and reliable medical device designed to capture, access and document a full set of patient vital signs that include temperature, blood pressure, pulse rate and SpO2 with a simple touchscreen display in about one minute. It allows hospitals, clinics or medical facilities to provide fast and accurate measurements right from the home screen.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.