Learn More
Labconco™ SpillStopper™ Work Surfaces for 8 Ft. Protector™Premier™and Protector™XL Hoods
SDPSCI_ED
SpillStopper Work Surfaces are molded of corrosion-resistant solid epoxy. They are contoured to conform to the interior liner of Protector Premier and XL Hoods.
$3927.00 - $4385.00
Specifications
For Use With (Application) 8 ft. Protector Premier and Protector XL Hoods
View More Specs
Products 7
Catalog Number Mfr. No. Product Type Price Quantity & Availability  
Catalog Number Mfr. No. Product Type Price Quantity & Availability  
S28244
SDPSCI_ED
missing translation for 'viewDocuments'
Labconco™
9500822/EMD
Left, Right Side (Cupsink Cutouts)
Each for $4,385.00
Add to cart
S28243
SDPSCI_ED
missing translation for 'viewDocuments'
Labconco™
9500811/EMD
Left, Right Rear (Cupsink Cutouts)
Each for $4,385.00
Add to cart
S28247
SDPSCI_ED
missing translation for 'viewDocuments'
Labconco™
9500820/EMD
Left Side (Cupsink Cutout)
Each for $4,185.00
Add to cart
S28245
SDPSCI_ED
missing translation for 'viewDocuments'
Labconco™
9500801/EMD
Right Rear (Cupsink Cutout)
Each for $4,185.00
Add to cart
S28246
SDPSCI_ED
missing translation for 'viewDocuments'
Labconco™
9500800/EMD
No cupsink cutout
Each for $3,927.00
Add to cart
S28248
SDPSCI_ED
missing translation for 'viewDocuments'
Labconco™
9500802/EMD
Right Side (Cupsink Cutout)
Each for $4,185.00
Add to cart
S28249
SDPSCI_ED
missing translation for 'viewDocuments'
Labconco™
9500810/EMD
Left Rear (Cupsink Cutout)
Each for $4,185.00
Add to cart
Description
Description
  • Molded from a special formulation of corrosion-resistant epoxy resin
  • Dished surface to contain spills
  • Contoured to conform to the interior liner of Labconco Protector Laboratory Hoods
  • Front edge has a large radius to aerodynamically direct airflow into the hood
  • Pre-drilled 2.5 in. (6.4cm) diameter holes for plumbing pass-through
  • Pre-drilled 1.5 in. (5.8cm) diameter holes for venting
  • May be ordered with pre-cut 6 x 3 in. (15.2 x 7.6cm) oval cupsink cutout(s) in left rear, right rear, left side, or right side. Cupsink is sold separately
Order Info
Service fixture and cup sink sold separately
Specifications
Specifications
8 ft. Protector Premier and Protector XL Hoods