Learn More
Labconco™ SpillStopper™ Work Surfaces for 8 Ft. Protector™Premier™and Protector™XL Hoods
SDPSCI_ED
SpillStopper Work Surfaces are molded of corrosion-resistant solid epoxy. They are contoured to conform to the interior liner of Protector Premier and XL Hoods.
Supplier:  Labconco™ 9500811/EMD
 View more versions of this product
Catalog No. S28243
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
View alternate product
Description
Description
  • Molded from a special formulation of corrosion-resistant epoxy resin
  • Dished surface to contain spills
  • Contoured to conform to the interior liner of Labconco Protector Laboratory Hoods
  • Front edge has a large radius to aerodynamically direct airflow into the hood
  • Pre-drilled 2.5 in. (6.4cm) diameter holes for plumbing pass-through
  • Pre-drilled 1.5 in. (5.8cm) diameter holes for venting
  • May be ordered with pre-cut 6 x 3 in. (15.2 x 7.6cm) oval cupsink cutout(s) in left rear, right rear, left side, or right side. Cupsink is sold separately
Order Info
Service fixture and cup sink sold separately
Specifications
Specifications
Left, Right Rear (Cupsink Cutouts)
8 ft. Protector Premier and Protector XL Hoods
Product Suggestions
Product Suggestions