Learn More
Labconco™ SpillStopper™ Work Surfaces for 4ft. Protector™XStream Hoods
SDPSCI_ED
SpillStopper Work Surfaces are molded of corrosion-resistant solid epoxy. They are contoured to conform to the interior liner of Protector XStream Hoods.
$2510.00 - $2983.00
Specifications
Material Epoxy Resin
View More Specs
Products 7
Catalog Number Mfr. No. Product Type Price Quantity & Availability  
Catalog Number Mfr. No. Product Type Price Quantity & Availability  
S28191
SDPSCI_ED
View Documents
Labconco™
9503400/EMD
No Cutouts
Each for $2,510.00
Add to cart
S28187
SDPSCI_ED
View Documents
Labconco™
9503401/EMD
Right Rear
Each for $2,743.00
Add to cart
S28189
SDPSCI_ED
View Documents
Labconco™
9503410/EMD
Left Rear
Each for $2,743.00
Add to cart
S28188
SDPSCI_ED
View Documents
Labconco™
9503402/EMD
Right Side
Each for $2,743.00
Add to cart
S28190
SDPSCI_ED
View Documents
Labconco™
9503420/EMD
Left Side
Each for $2,743.00
Add to cart
S28193
SDPSCI_ED
View Documents
Labconco™
9503422/EMD
Left Side, Right Rear
Each for $2,983.00
Add to cart
S28192
SDPSCI_ED
View Documents
Labconco™
9503411/EMD
Left Rear, Right Rear
Each for $2,983.00
Add to cart
Description
Description
  • Molded from a special formulation of corrosion-resistant epoxy resin
  • Dished surface to contain spills
  • Contoured to conform to the interior liner of Labconco Protector Laboratory Hoods
  • Front edge has a large radius to aerodynamically direct airflow into the hood
  • Pre-drilled 2.5 in. (6.4cm) diameter holes for plumbing pass-through
  • Pre-drilled 1.5 in. (5.8cm) diameter holes for venting
  • May be ordered with pre-cut 6 x 3 in. (15.2 x 7.6cm) oval cupsink cutout(s) in left rear, right rear, left side, or right side. Cupsink is sold separately
Order Info
Service fixture and cup sink sold separately
Specifications
Specifications
Epoxy Resin