Learn More
Labconco™ SpillStopper™ Work Surfaces for 6 Ft. Protector™ Premier™ and Protector™ XL Hoods
SDPSCI_ED
SpillStopper Work Surfaces are molded of corrosion-resistant solid epoxy. They are contoured to conform to the interior liner of Protector Premier and XL Hoods.
$2963.00 - $3407.00
Specifications
Material Epoxy Resin
View More Specs
Products 7
Catalog Number Mfr. No. Product Type Price Quantity & Availability  
Catalog Number Mfr. No. Product Type Price Quantity & Availability  
S28236
SDPSCI_ED
View Documents
Labconco™
9500620/EMD
Left Side
Each for $3,175.00
Add to Cart
S28237
SDPSCI_ED
View Documents
Labconco™
9500601/EMD
Right Rear
Each for $3,175.00
Add to Cart
S28238
SDPSCI_ED
View Documents
Labconco™
9500610/EMD
Left Rear
Each for $3,175.00
Add to Cart
S28239
SDPSCI_ED
View Documents
Labconco™
9500600/EMD
No Cutouts
Each for $2,963.00
Add to Cart
S28241
SDPSCI_ED
View Documents
Labconco™
9500611/EMD
Left Rear, Right Rear
Each for $3,407.00
Add to Cart
S28240
SDPSCI_ED
View Documents
Labconco™
9500602/EMD
Right Side
Each for $3,175.00
Add to Cart
S28242
SDPSCI_ED
View Documents
Labconco™
9500622/EMD
Left Side, Right Rear
Each for $3,407.00
Add to Cart
Description
Description
  • Molded from a special formulation of corrosion-resistant epoxy resin
  • Dished surface to contain spills
  • Contoured to conform to the interior liner of Labconco Protector Laboratory Hoods
  • Front edge has a large radius to aerodynamically direct airflow into the hood
  • Pre-drilled 2.5 in. (6.4cm) diameter holes for plumbing pass-through
  • Pre-drilled 1.5 in. (5.8cm) diameter holes for venting
  • May be ordered with pre-cut 6 x 3 in. (15.2 x 7.6cm) oval cupsink cutout(s) in left rear, right rear, left side, or right side. Cupsink is sold separately
Order Info
Service fixture and cup sink sold separately
Specifications
Specifications
Epoxy Resin