Learn More
Labconco™ SpillStopper™ Work Surfaces for 8 ft. Protector™XStream Hoods
SDPSCI_ED
SpillStopper Work Surfaces are molded of corrosion-resistant solid epoxy. They are contoured to conform to the interior liner of Protector XStream Hoods.
Supplier:  Labconco™ 9503820/EMD
 View more versions of this product
Catalog No. S28210
Add to Cart
Description
Description
  • Molded from a special formulation of corrosion-resistant epoxy resin
  • Dished surface to contain spills
  • Contoured to conform to the interior liner of Labconco Protector Laboratory Hoods
  • Front edge has a large radius to aerodynamically direct airflow into the hood
  • Pre-drilled 2.5 in. (6.4cm) diameter holes for plumbing pass-through
  • Pre-drilled 1.5 in. (5.8cm) diameter holes for venting
  • May be ordered with pre-cut 6 x 3 in. (15.2 x 7.6cm) oval cupsink cutout(s) in left rear, right rear, left side, or right side. Cupsink is sold separately
Order Info
Service fixture and cup sink sold separately
Specifications
Specifications
1.25 in.
113 Kg
96 in.
Left Side Cupsink Cutout. Not for use with Protector Solvent Storage Cabinets.
243.8 cm
250 lb.
36 in.
3.2 cm
91.4 cm
8 ft. SpillStopper Work Surface