Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FyterTech Nonwovens Spilfyter™ Granular Sorbents
Designed for efficient cleaning of oils and grease
Supplier: FyterTech Nonwovens 888002
Description
- All-purpose oil and grease absorbent
- 1:1 weight to weight absorbency ratio
- Components: Paper fiber; Kaolin; Calcium carbonate; Titanium dioxide
Order Info
Packaged in 14kg (30 lb.) bag
Specifications
Sorbent | |
Efficient cleaning of oils and grease | |
14 kg |
Granular | |
30 lb. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction