Learn More
SPEX CertiPrep™ OdorEroder
Effectively neutralizes a wide range of chemical odors in the lab
Supplier:  SPEX CertiPrep™ ODER250G
 View more versions of this product
Catalog No. ODER250G
Item Discontinued Due to product restrictions, please to purchase or view availability for this product.This item has been discontinued by the supplier.
Description
Description
  • Non-toxic, environmentally safe purple beads trap odors, and convert them into harmless compounds that remain trapped in the beads
  • As odors get trapped, the beads start turning brown
Specifications
Specifications
Effectively neutralizes a wide range of chemical odors in the lab
250 g
Product Suggestions
Product Suggestions