Learn More
SPEX CertiPrep™ OdorEroder
Effectively neutralizes a wide range of chemical odors in the lab
Supplier: SPEX CertiPrep™ ODER100G
Description
- Non-toxic, environmentally safe purple beads trap odors, and convert them into harmless compounds that remain trapped in the beads
- As odors get trapped, the beads start turning brown
Specifications
Effectively neutralizes a wide range of chemical odors in the lab | |
100 g |