Learn More
Honeywell™ Sperian™ Survivair Max T-Series Mask Respirator
Economically-designed full-face respirator provides comfort and visibility.
Supplier:  Honeywell™ 852000H5
Catalog No. 19-037-547B
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. View substitute products »
Description
Description
An economical, low-maintenance respirator featuring the same optically correct lens with panoramic view, double-sealing silicone facepiece flange, and comfortable oral/nasal mask as the 8600 Series. Provides cost-effective performance and comfort with high visibility. Five-strap suspension offers a secure, balanced fit.
Order Info
Available in three sizes. Compatible with the complete line of Willson T-Series cartridges and filters.
Specifications
Specifications
NIOSH 42 CFR Part 84
Silicone
Medium
852000-H5
Bayonet
Full Facepiece Respirator
North™ N-Series line of cartridges, filters, and cartridge/filter combinations
5-point