Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
3M™ Speedglas™ Welding Lens 9100 Replacement Lens
Replacement lens only (Welding filter adaptor not included)
$17.00
Specifications
Color | Black |
---|---|
For Use With (Equipment) | 9100 Series Welding Helmets |
Product Type | Welding Lens |
Quantity | 2/Cs. |
Catalog Number | Mfr. No. | Lens Tint | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Lens Tint | Price | Quantity & Availability | |||||
19-071-115
|
3M™
7010385032 |
Passive shade 10 |
Case of 2 for $17.00
|
|
|||||
19-071-116
|
3M™
7010316592 |
Passive shade 11 |
Case of 2 for $17.00
|
|
|||||
19-071-117
|
3M™
7010385033 |
Passive shade 12 |
Case of 2 for $17.00
|
|
|||||
19-071-118
|
3M™
7010342412 |
Passive shade 13 |
Case of 2 for $17.00
|
|
|||||
Description
- 3M Catalog# 06-0005-P10, 06-0005-P11, 06-0005-P12, 06-0005-P13
Specifications
Black | |
9100 Series Welding Helmets | |
Welding Lens | |
2/Cs. |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title