Learn More
Spectroline™ UV Absorbing Face Shields
Click to view available options
Face Shield
Description
- Improve contrast between the fluorescent area and the background by eliminating the “blue haze” interference
- Face shield (UVF–80) provide maximum protection from extended or high intensity UV exposure and adjust to fit the face
Compliance
Face shield meets ANSI specifications and OSHA standards.
Specifications
Product Type | UV-absorbing Faceshield |