Learn More
Spectroline™ Camera Adapters for UV Cabinets
Snap-on adapters replace eyepiece on UV cabinets
Catalog Number | Mfr. No. | Description | Price | Qty | Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Description | Price | Qty | Availability | ||||
FB-CA-35
|
Spectroline™
CA35 |
Camera Adapter | Camera Adapter | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | ||||
FB-CA-DS34
|
Spectroline™
CADS34 |
8 x 10 in. field | 8 x 10 in. field | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | ||||
FB-CA-DS34M
|
Spectroline™
CADS34/M |
4 x 5 in. field | 4 x 5 in. field | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | ||||
Description
- Camera snaps onto adapter so that lens is perpendicular to plane of gel or TLC plate to give undistorted photograph of experimental results
- Fit Spectronics Corporation Photodocumentation Camera