Learn More
Randox Randox Liquid Protein Calibrators / Protein Linearity Set
SDP
Supplier:  Randox IT2691
Encompass_Preferred
Specific Protein Calibrator. This new material is liquid ready to use (2-8*C), has open vial stability of 30 days, & all values are traceable to Reference Standards. It can be used for Linearity Verification of 13 specific protein analytes such as: Anti Streptolysin O (ASO), Ceruloplasmin, Complement C3, Complement C4, CRP, Ferritin, Haptoglobin, IgA, IgG, IgM, Prealbumin, Rheumatoid Factor (RF), Transferrin
Catalog No. NC0639138
Due to product restrictions, please to purchase or view availability for this product.