Learn More
Ace Glass, Inc. 25ML SPEC GRAV BTL W/STOPPER
Supplier: Ace Glass, Inc. 542024
Guy-Lussac type 25mL specific gravity bottle with 10/18 stopper, unadjusted for calibration in the laboratory. When adjusted, the bottle is suitable for ASTM D 369.