Learn More
Newcomer Supply Specimen Chuck w/Colored O-ring & Orientation Mark, Round, 30 mm, Blue
Supplier: Newcomer Supply 6865B
The 30 mm specimen chuck has a colored o-ring and oriention mark. It has proper tolerance and soft aluminum. It is suitable for Leica (except new CM1950), TBS, Tanner, Avantik and other common cryostats. Available in Blue, Green, Orange, Pink, Dark Purple, Red, White and Yellow.