Learn More
Fine Science Tools Absorption Triangles - Unmounted, 20 mm, 500/pk
Supplier: Fine Science Tools 1810503
Product: Absorption Triangles - Unmounted/Sterile/ Material : Synthetic/ Length : 20mm/ Mounted : No/ Sterile: No/ Autoclave Safe: / Unit : 500