Learn More
Bel-Art™ SP Scienceware™ Polyethylene Utility Bags
SCI_ED
Reusable, transparent bags are well-suited for shipping or storage and are especially useful as sample bags
Supplier:  Bel-Art™ H131780810/EMD
 View more versions of this product
Catalog No. S42892
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
View alternate product
Description
Description
  • FDA approved material; Latex free
  • Different sizes available to best match particular packaging requirements
  • Not autoclavable
Specifications
Specifications
Polyethylene
10.5 x 8 in. (267 x 203mm)
267 mm
203 mm
0.102 mm
10.5 in.
8 in.
Product Suggestions
Product Suggestions