Learn More
Bel-Art™ SP Scienceware™ Polyethylene Drying Tubes with Tubing Fittings
Fill these unbreakable polyethylene straight tubes with a desiccant. Tubes supplied with fittings that accept 0.25 - 0.375 in. tubing.
Supplier: Bel-Art™ F199650000/EMD
Description
- Dimensions: 16mm I.D. x 19mm O.D. (0.675 x 0.75 in.) in three length options (4, 6 or 8 in.)
- Each tube is supplied with two polypropylene fittings which accept 6.4 to 9.5mm (0.25 to 0.375 in.) I.D. tubing
- Additional fitting tips can be purchased separately
- Drying agent not included
Specifications
Replacement Tube Tips | |
144 | |
22 mm | |
Polyethylene |
0.87 in. | |
6.4 to 9.5 mm | |
22 mm |