Learn More
Bel-Art™ SP Scienceware™ Lab-Aire™ II Single-Sided Electric Benchtop Glassware Dryers
SCI_ED
Supplier:  Bel-Art™ F188192301/EMD
 View more versions of this product
Catalog No. S43295
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
View alternate product
Description
Description
Lab-Aire™ II Single-Sided Electric Benchtop Glassware Dryers accelerate drying of labware with warm, circulating air while their compact footprint uses minimal benchtop space.
Conserve benchtop space by placing single-sided models up against the wall.
  • 400w of drying power gently circulates heated air into and around items to decreases drying time
  • Heat selector knob with three temperature settings: high, medium and low
  • Power button illuminates when in use
  • 8 Amp at 120V, circuit breaker and independent thermo-fuse protects unit from overheating
  • Efficiency is enhanced with replaceable polyurethane foam filter
  • Redirect air flow by inserting plugs instead of pegs into unused peg holes
  • Sturdy bases with excellent balance; can be placed on any flat surface
  • Removable polypropylene baskets for storage of small labware and extra pegs
  • 120V
  • Two or three tier option
  • Baskets hold small glassware including test tubes, pipets
Specifications
Specifications
ABS
2 Tier
16.75 x 7.5 x 15.7 in. (42.5 x 19 x 40cm)
40 cm
42.5 cm
19 cm
Single Sided Electric Dryer
2 Baskets, 19 pegs
15.7 in.
16.75 in.
7.5 in.
120 V
Product Suggestions
Product Suggestions