Learn More
CTL Scientific Supply Corp SP IND PH 6.4-8,PK/6 RL X16
SDP
Supplier:  CTL Scientific Supply Corp 902106
Encompass_Preferred
Special Indicator pH paper 6.4-8.0, pack of 6 rolls (each 5 meters length) in dispensers with color charts. Gradation of scale: <6.4 - 6.4 - 6.6 - 6.8 - 7.0 - 7.2 - 7.4 - 7.6 - 7.8 - 8.0 - >8.0
Catalog No. 50-117-3254