Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ SP 141
High affinity MDM2 inhibitor
$299.28 - $1248.16
Specifications
Purity | >98% |
---|---|
Description | SP 141 |
Molecular Weight (g/mol) | 324.38 |
Product Type | SP 141 |
Description
High affinity MDM2 inhibitor (Ki = 28 nM); directly binds MDM2. Inhibits MDM2 expression and induces MDM2 autoubiquitination and proteasomal degradation. Attenuates growth of breast cancer xenograft tumors. Cell permeable.Specifications
>98% | |
SP 141 | |
324.38 | |
SP 141 |
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title