Learn More
SouthernBiotech™ Protein G
Source E. coli
Supplier: SouthernBiotech™ 740601S
Description
- Protein G binds with the Fc portion if IgGs from many species including all human, and mouse (IgG1, weakly) subclasses
- Does not bind Igm, IgA, or IgE or serum albumin
Protein G: Affinity chromatography, IP
Protein G-HRP Conjugate: EIA, IHC
Specifications
Protein G-UNLB | |
Lyophilized | |
-20°C |
1 mg | |
Affinity Chromatography, gamma Immunoglobulin Capture, Immunoprecipitation |