Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fisherbrand™ Solvent-Vented™ and Safety-Labeled Wash Bottles
missing translation for 'orderingAttributeHoverText'
Chemical Label:
Acetone
Ethanol
Isopropanol
Methanol
Unlabeled
Description
- Wide mouth 2 in. (53mm) bottle for easy refilling
- Polypropylene closure with leakproof design, color-coded to aid in easy identification
- Translucent, LDPE solvent-resistant; Sodium Hypochlorite is white LDPE for light-protection between 230 and 450nm; Toluene is red LDPE bottle
- Provides compliance with OSHA requirements outlined in Hazard Communication Standard 29 CFR 1910.1200
- Labeled with six of the most commonly used solvent names or label your own; custom labeling available
Use LYOB only with solvents compatible with LDPE.
Specifications
Closure Material | Polypropylene |
Closure Size | 53 mm |
Chemical Label | Acetone |
Capacity | 16 oz., 500mL |
Capacity (Metric) | 500 mL |
Capacity (English) | 16 oz. |
Color | Translucent |
Material | LDPE |
Closure Color | Red |
Certifications/Compliance | OSHA 29 CFR 1910.1200 |
Show More |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction