Learn More
MilliporeSigma™ Supelco™ Solvent Reservoir for Threaded ST/NS Joints, Joint Size: ST/NS 45/40
Flash chromatography is useful for rapid, preparative separations with moderate resolutions.
$326.00 - $342.00
Specifications
Column Type Flash Chromatography
For Use With (Application) Flash Chromatography
Joint Size ST/NS 45/40
View More Specs
Products 2
Catalog Number Mfr. No. Capacity (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Capacity (Metric) Price Quantity & Availability  
11-100-9510
View Documents
MilliporeSigma™ Supelco™
Z2024441EA
1000 mL
Each for $326.00
Add to cart
11-100-9455
View Documents
MilliporeSigma™ Supelco™
Z2024601EA
2000 mL
Each for $342.00
Add to cart
Description
Description
Columns can accommodate any reservoir with same ST/NS: joint size.
Specifications
Specifications
Flash Chromatography
Flash Chromatography
ST/NS 45/40