Learn More
MilliporeSigma™ Supelco™ Solvent Reservoir with Ball Joints, Joint Size: SJ 65/40
Flash chromatography is useful for rapid, preparative separations with moderate resolutions.
Supplier:  MilliporeSigma™ Supelco™ Z1212741EA
Catalog No. 11-100-9693
Add to Cart
Description
Description
Requires second pinch clamp.
Specifications
Specifications
Flash Chromatography
Flash Chromatography
500 mL
SJ 65/40