Learn More
Solid Glass Beads
SCI_ED
Increase the efficiency of separations.
Supplier:  Walter Stern Inc. 100E/EMD
 View more versions of this product
Catalog No. S800241
Edge
Add to Cart
Description
Description
  • For use in distillations or as inert boiling stones
  • Packaged in 1 lb. (0.45kg) packs
Specifications
Specifications
Glass
4 mm
5500/Pk.
Product Suggestions
Product Suggestions