Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Sodium L-Lactate Solution, USP, 60%, Spectrum™ Chemical
Click to view available options
Quantity:
500 mL
4 L
20 L
200 L
Packaging:
Amber Glass Bottle
Drum
Poly Drum
Description
Spectrum™ Chemical Sodium L-Lactate Solution, 60 Percent, USP is used for replacing fluids and electrolytes in those who have low blood volume or low blood pressure. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Specifications
pH | 5.0 to 9.0 |
CAS Max % | 0.6, 0.4 |
Percent Purity | 58.8 to 61.2% |
Grade | USP |
Packaging | Drum |
Quantity | 200 L |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction