Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Sodium Hydroxide Solution
SCI_ED

Click to view available options
Quantity:
500 mL
Packaging:
0.2M
0.5M
3M
50%
6M
1310-73-2, 7732-18-5
HNaO
39.997
HEMHJVSKTPXQMS-UHFFFAOYSA-M
sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic
14798
CHEBI:32145
sodium;hydroxide
[OH-].[Na+]
1310-73-2, 7732-18-5
HNaO
39.997
HEMHJVSKTPXQMS-UHFFFAOYSA-M
sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic
14798
CHEBI:32145
sodium;hydroxide
[OH-].[Na+]

This product is for educational use - no C of A is available.

Density 1.000
Quantity 500 mL
Packaging 3M
Physical Form Liquid
Chemical Name or Material Sodium Hydroxide Solution

ShelfLife : 2 years

DOTInformation : DOT Class 8, II, : Corrosive