Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Hydroxide, 0.02 N Solution, APHA, Spectrum™ Chemical
SDP

Click to view available options
Quantity:
500 mL
1 L
4 L
20 L
Packaging:
Cubitainer
Poly Bottle
1310-73-2, 7732-18-5
1310-73-2, 7732-18-5
Concentration 0.02 N ± 0.0005 N
Quantity 20 L
Grade Chemical Solution
Packaging Cubitainer

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.