Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Sodium, Fisher Chemical
Click to view available options
Quantity:
1 lb.
Packaging:
Metal Can
Specifications
Color | Gray |
Melting Point | 98°C |
Atomic Weight | 22.99 |
Quantity | 1 lb. |
Identification | Pass Test |
Formula Weight | 22.99 |
Packaging | Metal Can |
Vapor Pressure | 1mmHg at 440°C |
Form | Solid |
Chemical Name or Material | Sodium |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction