missing translation for 'onlineSavingsMsg'
Learn More

Fisher Science Education™ Sodium Bromide
SCI_ED

Catalog Number S25538A
missing translation for 'orderingAttributeHoverText'
Quantity:
100 g
500 g
Packaging:
Reagent Grade
7647-15-6
BrNa
102.89
MFCD00003475
JHJLBTNAGRQEKS-UHFFFAOYSA-M
sodium bromide, bromide salt of sodium, sodium bromide nabr, sedoneural, sodiumbromide, trisodium tribromide, bromnatrium, nabr, bromnatrium german, caswell no. 750a
253881
CHEBI:63004
sodium bromide
[Na+].[Br-]
7647-15-6
BrNa
102.89
MFCD00003475
JHJLBTNAGRQEKS-UHFFFAOYSA-M
sodium bromide, bromide salt of sodium, sodium bromide nabr, sedoneural, sodiumbromide, trisodium tribromide, bromnatrium, nabr, bromnatrium german, caswell no. 750a
253881
CHEBI:63004
sodium bromide
[Na+].[Br-]

This product is for educational use - no C of A is available.

Color White
Quantity 500 g
Assay Percent Range >99%
Linear Formula NaBr
Formula Weight 102.9
Grade Reagent
Packaging Reagent Grade
Density 3.208g/mL
Chemical Name or Material Sodium Bromide
ShelfLife 1 year
missing translation for 'provideContentCorrection'

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

missing translation for 'productTitle'