Learn More
SNX1 293T Cell Overexpression Lysate 1 (Denatured), Abnova
Supplier:  Abnova Corporation H00006642T01
Catalog No. 89-000-012
Add to Cart
Description
Description
Western Blotting
Specifications
Specifications
293T
SNX1 293T Cell Transient Overexpression Lysate(Denatured)
59.1
pCMV-SNX1 full-length
100 μL
Human
Western Blot
NM_003099
6.8
Transient overexpression cell Lysate was tested with Anti-SNX1 antibody (H00006642-B01) by western blotting
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue)
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Product Suggestions
Product Suggestions