Learn More
Smithsonian Institution™ Weather Center
Supplier: Neo/Sci Corporation 203573
Description
Gain a basic understanding of key weather factors such as temperature, total rainfall, wind speed, and wind direction.
• Complete weather tower storage system is self-contained and can be mounted outdoors on any fence or rail
• Includes anemometer, wind vane, compass, indoor and outdoor thermometers, rain gauge, data log, cloud charts, and weather tower storage case
• Detailed, fully illustrated literature features reference information on meteorology and step-by-step instructions
Order Info
Dimensions: 12L x 4W x 10 in. H.
Specifications
Anemometer, wind vane, compass, indoor and outdoor thermometers, rain gauge, data log, cloud charts, and weather tower storage case | |
Smithsonian Weather Class Set 5 | |
4 in. | |
Self contained and can be mounted outdoors on any fence or rail, gain a basic understanding of key weather factors such as temperature, total rainfall, wind speed, and wind direction |
10.1 cm | |
25.4 cm | |
10 in. |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.