Learn More
ICU Medical Portex™ Line Draw Plus Arterial Blood Sampling Kits
Line Draw Plus aspirating syringes allow for blood sampling directly from an arterial line. The syringes use an advanced formula of dry lithium heparin for consistent and accurate readings. They are available with the following configurations: 1 mL Luer Slip Syringe, 3 mL Luer Lock Syringe, or 3 mL Luer Slip Slip Syringe.
Supplier: ICU Medical 40532
Description
Advanced formula offsets both binding and dilution factors by artificially pre-adjusting each electrolyte and metabolite to its normal blood level. Provides consistent and accurate readings for pH, pCO2, pO2, Ca2+ , Mg2+ , Na+, K+, Cl-, CO-oximetry, glucose, and lactate.
- Umbilical and arterial syringes contain a 23.5 IU/mL concentration of dry lithium heparin
- Low-heparin arterial syringes have a 7 IU/mL concentration of dry lithium heparin
- Anaerobic syringes contain 23.5 IU/mL of electrolyte-balanced liquid sodium heparin
Specifications
Allow for blood sampling directly from an arterial line, provides consistent and accurate analysis for all parameters tested | |
Arterial Blood Sampling Kit | |
Polybag |
1mL line draw luer-slip syringe with Filter-Pro™ device, 23.5 IU/mL of electrolyte-balanced anaerobic liquid sodium heparin | |
200/Cs. | |
3 mL |
Safety and Handling
ShelfLife : 3 Years
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.