Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Smiths Medical Needle-Pro™ Needle with Needle Protection Device

Ideal for clinical applications like intramuscular and subcutaneous injections

Supplier:  Smiths Medical 4293

Catalog No. 22-004-268



Due to product restrictions, please to purchase or view availability for this product.


Description

Description

  • For use with Luer slip syringes
  • Needle is securely locked inside the safety device and remains protective through disposal
  • Needle hub color is orange
  • Needle gauge: 25 G

Order Info

Length: 1.5 in. (3.81cm)

Compliance

Meets NIOSH/CDC recommendations

Specifications

Specifications

25 G
Rotating Needle-Pro device
Hypodermic Needle
Ideal for clinical applications like intramuscular and subcutaneous injections, luer-slip syringes
1.5 in.
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit