Learn More
ICU Medical Needle-Pro™ EDGE™ Fixed Needle Insulin Syringe
Single-handed technique
Supplier: ICU Medical 44281
Description
The needle is securely locked inside the safety device and remains protective through disposal.
- Available with 28 and 29 G needles fixed to 0.3, 0.5 and 1.0mL U-100 insulin syringes
Specifications
NIOSH/CDC | |
Hypodermic fixed needle | |
1.27 cm | |
1 mL |
28 G | |
0.5 in. | |
Insulin Syringe |