Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Wanzl SmartCell™ Chrome Shelving Divider

Chrome Plus Basket Divider for SmartCell Shelving System.

$17.00 - $30.35

Specifications

Color Chrome
For Use With (Equipment) SmartCell™
For Use With (Application) Shelving
Includes 1 Divider
Material Steel
View More Specs

Products 4
Catalog Number Mfr. No. Dimensions (L x W) Price Quantity & Availability  
Catalog Number Mfr. No. Dimensions (L x W) Price Quantity & Availability  
11-222-027
View Documents
Wanzl
T.H36DV
3 x 6 in.
Each for $17.00
Add to Cart
 
11-222-028
View Documents
Wanzl
T.H66DV
6 x 6 in.
Each for $21.15
Add to Cart
 
11-222-029
View Documents
Wanzl
T.H126DV
12 x 6 in.
Each for $27.00
Add to Cart
 
11-222-030
View Documents
Wanzl
T.H186DV
18 x 6 in.
Each for $30.35
Add to Cart
 
Description

Description

  • Allow for custom compartmentalization of the SmartCell™ basket system.
  • Large and small articles alike are stored in separate compartments, allowing for easy identification and quick retrieval
Specifications

Specifications

Chrome
Shelving
Steel
SmartCell™
1 Divider
6 in.
SDS
Documents

Documents

Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit