Learn More
USA BLUE BOOK Sludge Judge II Complete Set
Supplier: USA BLUE BOOK 25513
Sludge Judge samplers take accurate measurements of settleable solids in slurries that contain less than 5% solids. All units consist of three 5-ft sections for easy transport. Sludge Judge II features a 1-1/4" butyrate tube. That’s double the size of the original Sludge Judge‚ resulting in double the capacity—6 oz per ft.