Learn More
Thomas Scientific Sludge Judge, 15 ft.
Supplier: Thomas Scientific 8257M32
The Sludge Judge is designed to take accurate readings of settleable solids, 5% or less, in a variety of liquids, to any depth. It is ideal for sewage treatment plants, chemical plants, and food processing facilities - anywhere that accurate sample levels of settleable solids in noncaustic materials are needed. The unit holds approximately 3 oz. per foot (89 ml per 0.31 m). The Sludge Judge comes in 5 ft. (1.53 m) sections of 3/4" (1.90 cm) plastic pipe with screw-type connectors. The top section includes a nylon rope for raising and lowering the sampler. Individual sections can be combined as required to achieve the length needed. Do not use in liquids over 165°F (74°C). Sludge Judge is not autoclavable. A convenient cotton canvas carrying case that holds four sections, a cleaning brush, cleaning rod, and cotton strips are available. To use: Slowly lower the Sludge Judge to the bottom of the tank. The float valve opens, allowing materials to flow in. W
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.