Learn More
Simport™ Scientific Slimsette™ Tissue Processing/Embedding Cassettes in Quickload™ Stacks
Secure specimens during processing with Slimsette™ Tissue Processing/Embedding Cassettes in Quickload™ Stacks.
$1157.00 - $1193.00
Specifications
Quantity 2000/Pk.
Material Acetal
View More Specs
Products 11
Catalog Number Mfr. No. Color Price Quantity & Availability  
Catalog Number Mfr. No. Color Price Quantity & Availability  
22-038-529
View Documents
Simport™ Scientific
M5092T
White
Case of 2000 for $1,157.00
Add to Cart
22-038-530
View Documents
Simport™ Scientific
M5093T
Pink
Case of 2000 for $1,157.00
Add to Cart
22-038-531
View Documents
Simport™ Scientific
M5094T
Green
Case of 2000 for $1,193.00
Add to Cart
22-038-532
View Documents
Simport™ Scientific
M5095T
Yellow
Case of 2000 for $1,157.00
Add to Cart
22-038-533
View Documents
Simport™ Scientific
M5096T
Blue
Case of 2000 for $1,157.00
Add to Cart
22-038-534
View Documents
Simport™ Scientific
M5097T
Peach
Case of 2000 for $1,157.00
Add to Cart
22-038-535
View Documents
Simport™ Scientific
M5098T
Tan
Case of 2000 for $1,157.00
Add to Cart
22-038-536
View Documents
Simport™ Scientific
M5099T
Gray
Case of 2000 for $1,157.00
Add to Cart
22-038-537
View Documents
Simport™ Scientific
M50910T
Lilac
Case of 2000 for $1,157.00
Add to Cart
22-038-538
View Documents
Simport™ Scientific
M50911T
Orange
Case of 2000 for $1,157.00
Add to Cart
22-038-539
View Documents
Simport™ Scientific
M50912T
Aqua
Case of 2000 for $1,157.00
Add to Cart
Description
Description
  • In Quickload™ stacks
  • 45° Anterior writing area
  • Recessed cover
  • Suitable for automated printers
  • Made of acetal
  • Resistant to the chemical action of most histological solvents
Order Info
Each case contains 50 Quickload™ stacks of 40 cassettes.
Certifications
CE-IVD
Specifications
Specifications
2000/Pk.
Acetal